N,N-Dimethylallylamine, 98%
N,N-Dimethylallylamine, 98%
N,N-Dimethylallylamine, 98%
Thermo Scientific Chemicals

N,N-Dimethylallylamine, 98%

CAS: 2155-94-4 | C5H11N | 85.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 407980050
Price (USD)
81.65
Special offer
Online exclusive
Ends: 28-Dec-2026
96.30
Save 14.65
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
81.65
Special offer
Online exclusive
Ends: 28-Dec-2026
96.30
Save 14.65
Each
Chemical Identifiers
CAS2155-94-4
IUPAC Namedimethyl(prop-2-en-1-yl)amine
Molecular FormulaC5H11N
InChI KeyGBCKRQRXNXQQPW-UHFFFAOYSA-N
SMILESCN(C)CC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.4000 to 1.4020 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only