2,6-Dimethyl-3,5-heptanedione, 99%
2,6-Dimethyl-3,5-heptanedione, 99%
2,6-Dimethyl-3,5-heptanedione, 99%
Thermo Scientific Chemicals

2,6-Dimethyl-3,5-heptanedione, 99%

CAS: 18362-64-6 | C9H16O2 | 156.225 g/mol
数量:
5 g
25 g
パッケージ:
Glass Bottle
製品番号(カタログ番号) 408360250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS18362-64-6
IUPAC Name2,6-dimethylheptane-3,5-dione
Molecular FormulaC9H16O2
InChI KeyCEGGECULKVTYMM-UHFFFAOYSA-N
SMILESCC(C)C(=O)CC(=O)C(C)C
さらに表示
Appearance (Form)Liquid
Refractive index1.4560 to 1.4600 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only