N,N-Dimethyl-p-phenylenediamine oxalate, 99%
N,N-Dimethyl-p-phenylenediamine oxalate, 99%
N,N-Dimethyl-p-phenylenediamine oxalate, 99%
Thermo Scientific Chemicals

N,N-Dimethyl-p-phenylenediamine oxalate, 99%

CAS: 62778-12-5 | 0·5 C2H2O4 | 362.42 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 408490250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS62778-12-5
IUPAC Namebis(N1,N1-dimethylbenzene-1,4-diamine); oxalic acid
Molecular FormulaC18H26N4O4
InChI KeyMNUINXKPLPIOEF-UHFFFAOYSA-N
SMILESOC(=O)C(O)=O.CN(C)C1=CC=C(N)C=C1.CN(C)C1=CC=C(N)C=C1
さらに表示
Appearance (Form)Powder and/or chunks
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Color)Beige to grey to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only