N,N-Dimethyl-p-phenylenediamine oxalate, 99%
N,N-Dimethyl-p-phenylenediamine oxalate, 99%
N,N-Dimethyl-p-phenylenediamine oxalate, 99%
Thermo Scientific Chemicals

N,N-Dimethyl-p-phenylenediamine oxalate, 99%

CAS: 62778-12-5 | 0·5 C2H2O4 | 362.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 408491000
Price (USD)
432.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
432.00
Each
Chemical Identifiers
CAS62778-12-5
IUPAC Namebis(N1,N1-dimethylbenzene-1,4-diamine); oxalic acid
Molecular FormulaC18H26N4O4
InChI KeyMNUINXKPLPIOEF-UHFFFAOYSA-N
SMILESOC(=O)C(O)=O.CN(C)C1=CC=C(N)C=C1.CN(C)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or chunks
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Color)Beige to grey to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only