3,5-Dinitrobenzoyl Chloride, 99%
3,5-Dinitrobenzoyl Chloride, 99%
3,5-Dinitrobenzoyl Chloride, 99%
Thermo Scientific Chemicals

3,5-Dinitrobenzoyl Chloride, 99%

CAS: 99-33-2 | C7H3ClN2O5 | 230.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 408711000
Price (USD)
129.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
129.00
Each
Chemical Identifiers
CAS99-33-2
IUPAC Name3,5-dinitrobenzoyl chloride
Molecular FormulaC7H3ClN2O5
InChI KeyNNOHXABAQAGKRZ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(=CC(=C1)C(Cl)=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to beige
Titration RCOCl>=98.5 %
Appearance (Form)Crystals or crystalline needles
Infrared spectrumConforms
Melting point67°C to 71°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only