Dipropyl phthalate, 99+%
Dipropyl phthalate, 99+%
Dipropyl phthalate, 99+%
Thermo Scientific Chemicals

Dipropyl phthalate, 99+%

CAS: 131-16-8 | C14H18O4 | 250.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 409161000
Price (USD)
234.65
Online exclusive
261.00
Save 26.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
234.65
Online exclusive
261.00
Save 26.35
Each
Chemical Identifiers
CAS131-16-8
IUPAC Name1,2-dipropyl benzene-1,2-dicarboxylate
Molecular FormulaC14H18O4
InChI KeyMQHNKCZKNAJROC-UHFFFAOYSA-N
SMILESCCCOC(=O)C1=CC=CC=C1C(=O)OCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Refractive index1.4960 to 1.4980 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only