Dipropyl phthalate, 99+%
Dipropyl phthalate, 99+%
Dipropyl phthalate, 99+%
Thermo Scientific Chemicals

Dipropyl phthalate, 99+%

CAS: 131-16-8 | C14H18O4 | 250.29 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
產品號碼 409165000
價格 (TWD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS131-16-8
IUPAC Name1,2-dipropyl benzene-1,2-dicarboxylate
Molecular FormulaC14H18O4
InChI KeyMQHNKCZKNAJROC-UHFFFAOYSA-N
SMILESCCCOC(=O)C1=CC=CC=C1C(=O)OCCC
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Refractive index1.4960 to 1.4980 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only