Dodecyltrimethylammonium bromide, 99%, pure
Dodecyltrimethylammonium bromide, 99%, pure
Dodecyltrimethylammonium bromide, 99%, pure
Thermo Scientific Chemicals

Dodecyltrimethylammonium bromide, 99%, pure

CAS: 1119-94-4 | C15H34BrN | 308.34 g/mol
Quantity:
100 g
500 g
Catalog number 409315000
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS1119-94-4
IUPAC Namedodecyltrimethylazanium bromide
Molecular FormulaC15H34BrN
InChI KeyXJWSAJYUBXQQDR-UHFFFAOYSA-M
SMILES[Br-].CCCCCCCCCCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Color of solution=<100 APHA (10% aq. soln.)
Solubility(10 % in water) Clear to slightly hazy
Titration Argentometric>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only