Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%
Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%
Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%
Thermo Scientific Chemicals

Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%

CAS: 67-42-5 | C14H24N2O10 | 380.34 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 409915000
価格(JPY)
-
数量:
500 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS67-42-5
IUPAC Name3,12-bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid
Molecular FormulaC14H24N2O10
InChI KeyDEFVIWRASFVYLL-UHFFFAOYSA-N
SMILESOC(=O)CN(CCOCCOCCN(CC(O)=O)CC(O)=O)CC(O)=O
さらに表示
Titration excess NaOH>=97.5 %
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Crystalline powder
Melting point240°C to 244°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only