Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, ACS reagent
Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, ACS reagent
Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, ACS reagent
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, ACS reagent

CAS: 6381-92-6 | C10H18N2Na2O10 | 372.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 409970025
Price (USD)
552.65
Online exclusive
614.00
Save 61.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
552.65
Online exclusive
614.00
Save 61.35
Each
Chemical Identifiers
CAS6381-92-6
IUPAC Namedisodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular FormulaC10H18N2Na2O10
InChI KeyOVBJJZOQPCKUOR-UHFFFAOYSA-L
SMILESO.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
pH4.0 to 6.0 (5 % at 25°C)
Appearance (Color)White to light yellow
Titration Complexometric99.0 to 101.0 %
Insoluble matter=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only