LYVE1 Monoclonal Antibody (ALY7), eFluor™ 570, eBioscience™ - FAQs
LYVE1 Monoclonal Antibody (ALY7), eFluor™ 570, eBioscience™ - FAQs
View additional product information for LYVE1 Monoclonal Antibody (ALY7), eFluor™ 570, eBioscience™ - FAQs (41-0443-82, 41-0443-80)