FOXP3 Monoclonal Antibody (236A/E7), eFluor™ 570, eBioscience™ - FAQs
FOXP3 Monoclonal Antibody (236A/E7), eFluor™ 570, eBioscience™ - FAQs
View additional product information for FOXP3 Monoclonal Antibody (236A/E7), eFluor™ 570, eBioscience™ - FAQs (41-4777-80, 41-4777-82)