F4/80 Monoclonal Antibody (BM8), eFluor™ 570, eBioscience™ - FAQs
F4/80 Monoclonal Antibody (BM8), eFluor™ 570, eBioscience™ - FAQs
View additional product information for F4/80 Monoclonal Antibody (BM8), eFluor™ 570, eBioscience™ - FAQs (41-4801-82, 41-4801-80)