OCT3/4 Monoclonal Antibody (EM92), eFluor™ 570, eBioscience™ - FAQs
OCT3/4 Monoclonal Antibody (EM92), eFluor™ 570, eBioscience™ - FAQs
View additional product information for OCT3/4 Monoclonal Antibody (EM92), eFluor™ 570, eBioscience™ - FAQs (41-5841-82, 41-5841-80)