2-Ethylhexanal, 96%
2-Ethylhexanal, 96%
2-Ethylhexanal, 96%
Thermo Scientific Chemicals

2-Ethylhexanal, 96%

CAS: 123-05-7 | C8H16O | 128.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 410090010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS123-05-7
IUPAC Name2-ethylhexanal
Molecular FormulaC8H16O
InChI KeyLGYNIFWIKSEESD-UHFFFAOYNA-N
SMILESCCCCC(CC)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4125 to 1.4185 (20°C, 589 nm)
Free acid=<2.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only