Hypersil™ PREP HS C18 Bulk Media
Actual product may vary
Thermo Scientific™
Hypersil™ PREP HS C18 Bulk Media
Ensure optimal chromatography performance and cost savings with Hypersil PREP HS C18 bulk chromatography media.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
41010-01000B1000 g
41010-00050B50 g
41010-00100B100 g
41010-00500B500 g
41010-05000B5000 g
41010-10000B10000 g
Catalog number 41010-01000B
Price (USD)
-
Quantity:
1000 g
Ensure optimal chromatography performance with our Thermo Scientific™ Hypersil™ PREP HS C18 bulk chromatography media. Purchase bulk quantities of silica-based, high surface area C18 media to repack preparative HPLC columns yourself without paying for new hardware.
Repack Columns Yourself with Bulk Media
Excellent Choice for Routine Applications
  • C18 (USP L1) selectivity.
  • High surface area particles with 100Å pore size.
  • pH stable from 2 to 9.
  • Maximum temperature of 60°C.
Customer Specific Orders
  • Please contact your local sales representative for Customer Specific Orders.
Specifications
Carbon Load15%
Column TypeReversed Phase
EndcappedYes
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
PhaseReversed Phase
Pore Size100 Å
Product LinePrep LC
Quantity1000 g
Stationary PhaseC18
Surface Area (Metric)350 m²/g
Temperature60°C
USP TypeL1
Particle Size10 μm
pH2 to 9
Unit SizeEach