2-Ethylhexyl acrylate, 99+%, stabilized
2-Ethylhexyl acrylate, 99+%, stabilized
2-Ethylhexyl acrylate, 99+%, stabilized
Thermo Scientific Chemicals

2-Ethylhexyl acrylate, 99+%, stabilized

CAS: 103-11-7 | C11H20O2 | 184.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 410110010
Price (USD)
48.65
Online exclusive
53.80
Save 5.15
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
48.65
Online exclusive
53.80
Save 5.15
Each
Chemical Identifiers
CAS103-11-7
IUPAC Name2-ethylhexyl prop-2-enoate
Molecular FormulaC11H20O2
InChI KeyGOXQRTZXKQZDDN-UHFFFAOYNA-N
SMILESCCCCC(CC)COC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.05 %
Infrared spectrumConforms
Stabilizer10 to 20 ppm MEHQ
Appearance (Form)Clear liquid
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only