Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0%, USP, GMP, Multi-Compendial, J.T.Baker™
Avantor™

Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0%, USP, GMP, Multi-Compendial, J.T.Baker™

This product is GMP Manufactured meets JPC (1997), BP, EP and USP Requirements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
4102-052.5 kgWhite Poly Bottle, Tamper-evident Cap
4102-0712 kgPolyethylene Pail, Screw Top
4102-F950 kgDrum, HDPE Flowmor Package
4102-0950 kgPolyethylene Drum, Open Head
4102-01500 gWhite Poly Bottle, Tamper-evident Cap
4102-F712 kgDrum, HDPE Flowmor Package
Catalog number 4102-05
Price (MXN)
-
Quantity:
2.5 kg
Packaging:
White Poly Bottle, Tamper-evident Cap
This product is GMP Manufactured meets JPC (1997), BP, EP and USP Requirements.
This product is GMP Manufactured meets JPC (1997), BP, EP and USP Requirements.
Specifications
Boiling Point357°C (1013 hPa)
CAS77-86-1
CAS77-86-1
Chemical Name or MaterialTris(hydroxymethyl)aminomethane
ColorColorless
Melting Point169–172°C
Molecular FormulaNH2C(CH2OH)3
PackagingWhite Poly Bottle, Tamper-evident Cap
Synonym2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris(hydroxymethyl)methylamine, TRIS base
For Use With (Application)Pharma and Biopharma
GradeUSP
Molecular Weight (g/mol)121.14
Percent Purity99.0-101.0%
Physical FormCrystalline
Quantity2.5 kg
Solution TypeBuffer
pH10.0-11.5
Unit SizeEach