TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), ≥99.0% (dried basis), Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker™
Avantor™

TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), ≥99.0% (dried basis), Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker™

For laboratory, research, or manufacturing use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
4103-0712 kgPolyethylene Pail, Screw Top
4103-01500 gWhite Poly Bottle, Tamper-evident Cap
4103-021 kgWhite Poly Bottle, Tamper-evident Cap
4103-04250 gWhite Poly Bottle, Tamper-evident Cap
4103-065 kgPolyethylene Pail, Screw Top
4103-0850 kgPolyethylene Drum, Open Head
4103-F712 kgPail, HDPE Flowmor Package
Catalog number 4103-07
Price (MXN)
-
Quantity:
12 kg
Packaging:
Polyethylene Pail, Screw Top
For laboratory, research, or manufacturing use.
For laboratory, research, or manufacturing use.
Specifications
CAS1185-53-1
Chemical Name or MaterialTRIS HCl
ColorColorless
Melting Point150–152°C
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingPolyethylene Pail, Screw Top
SynonymTris(hydroxymethyl)aminomethane hydrochloride, Tris(hydroxymethyl)methylamine hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)methylammonium chloride, THAM HCl
For Use With (Application)Pharma and Biopharma
GradeMolecular Biology Ultrapure
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
Physical FormCrystalline
Quantity12 kg
Solution TypeBuffer
Unit SizeEach