Ethyl benzoylformate, 95%
Ethyl benzoylformate, 95%
Ethyl benzoylformate, 95%
Thermo Scientific Chemicals

Ethyl benzoylformate, 95%

CAS: 1603-79-8 | C10H10O3 | 178.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 410350050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1603-79-8
IUPAC Nameethyl 2-oxo-2-phenylacetate
Molecular FormulaC10H10O3
InChI KeyQKLCQKPAECHXCQ-UHFFFAOYSA-N
SMILESCCOC(=O)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5150 to 1.5170 (20°C, 589 nm)
Appearance (Color)Clear colorless to green
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only