TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), ≥99.0%, from Tromethamine USP, GMP, Biotech Reagent, J.T.Baker™
Avantor™

TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), ≥99.0%, from Tromethamine USP, GMP, Biotech Reagent, J.T.Baker™

GMP Manufactured Product.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
4106-F712 kgPail, HDPE Flowmor Package
4106-01500 gWhite Poly Bottle, Tamper-evident Cap
4106-052.5 kgWhite Poly Bottle, Tamper-evident Cap
4106-0712 kgPolyethylene Pail, Screw Top
4106-0950 kgPolyethylene Drum, Open Head
4106-F950 kgDrum, HDPE Flowmor Package
Catalog number 4106-F7
Price (MXN)
-
Quantity:
12 kg
Packaging:
Pail, HDPE Flowmor Package
GMP Manufactured Product.
GMP Manufactured Product.
Specifications
CAS1185-53-1
Chemical Name or MaterialTRIS HCl
ColorColorless
Melting Point147–153°C
Molecular FormulaNH2C(CH2OH)3·HCl
PackagingPail, HDPE Flowmor Package
SynonymTRIS HCl THAM HCl Tris(hydroxymethyl)methylamine hydrochloride Trometamol hydrochloride Tris(hydroxymethyl)aminomethane hydrochloride 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride Tris(hydroxymethyl)methylammonium chloride
For Use With (Application)Pharma and Biopharma
GradeMolecular Biology Ultrapure
Molecular Weight (g/mol)157.6
Percent Purity≥99.0%
Physical FormCrystalline
Quantity12 kg
Solution TypeBuffer
Unit SizeEach