Fluorexon
Fluorexon
Fluorexon
Thermo Scientific Chemicals

Fluorexon

CAS: 1461-15-0 | C30H26N2O13 | 622.539 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 410630050
Price (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1461-15-0
IUPAC Name2-{[(7'-{[bis(carboxymethyl)amino]methyl}-3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-2'-yl)methyl](carboxymethyl)amino}acetic acid
Molecular FormulaC30H26N2O13
InChI KeyDEGAKNSWVGKMLS-UHFFFAOYSA-N
SMILESOC(=O)CN(CC(O)=O)CC1=CC2=C(OC3=CC(O)=C(CN(CC(O)=O)CC(O)=O)C=C3C22OC(=O)C3=CC=CC=C23)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Suitability(for Ca determination) Passes test
UV spectrumComplies
Appearance (Color)Yellow to orange-brown
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only