Fmoc-L-Isoleucine, 10/Package

Catalog number: 410640

Applied Biosystems™  Related applications: Protein Sequencing & Peptide Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

These are Amino Acid Cartridges for the ABI™ 431-433 Peptide Synthesizer.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 433A Peptide Synthesizer
Format: Cartridge
Product Line: Applied Biosystems™
Product Size: 10 each

Contents & storage

Each package contains 10 cartridges.

Documents

Manuals & protocols