Fluorescein, disodium salt dihydrate, 90%, pure
Fluorescein, disodium salt dihydrate, 90%, pure
Fluorescein, disodium salt dihydrate, 90%, pure
Thermo Scientific Chemicals

Fluorescein, disodium salt dihydrate, 90%, pure

CAS: 1264198-62-0 | C20H10Na2O5·2H2O
数量:
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 410641000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1264198-62-0
Appearance (Color)Orange-red to green-brown to dark brown
Infrared spectrumConforms
Water6.5 to 10 % (K.F.)
Appearance (Form)Powder
UV spectrophotometry>=88 % (on dry substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only