Nalgene™ PMP Erlenmeyer Flasks with Closure
Nalgene™ PMP Erlenmeyer Flasks with Closure
Nalgene™ PMP Erlenmeyer Flasks with Closure
Nalgene™ PMP Erlenmeyer Flasks with Closure
Thermo Scientific™

Nalgene™ PMP Erlenmeyer Flasks with Closure

Perform titrations, preparation and storage of culture media with these flasks, which provide a combination of chemical resistance, strength and transparency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
4109-1000PK
also known as 4109-1000
1000 mL53 mmPack of 2
4109-0125125 mL33 mmCase of 24
4109-0125PK125 mL33 mmPack of 6
4109-0250250 mL38 mmCase of 12
4109-0250PK
also known as 4109-0250
250 mL38 mmPack of 4
4109-0500500 mL43 mmCase of 12
4109-0500PK
also known as 4109-0500
500 mL43 mmPack of 4
4109-10001000 mL53 mmCase of 6
Catalog number 4109-1000PK
also known as 4109-1000
Price (EUR)
74,10
Pack of 2
Capacity (Metric):
1000 mL
Closure Size:
53 mm
Unit Size:
Pack of 2
Price (EUR)
74,10
Pack of 2

Perform titrations, preparation and storage of culture media with Thermo Scientific™ Nalgene™ PMP Erlenmeyer Flasks with Closure, which provide a combination of chemical resistance, strength and transparency. Rugged, shatter-proof construction combined with light weight make these flasks a valuble element of lab safety programs.

Features:
  • Excellent chemical resistance to a wide variety of lab reagents
  • More transparent than polypropylene but with similar chemical resistance
  • Ideal for preparation and storage of culture media and titrations
  • All sizes have easy-to-read graduations
  • Autoclavable/Graduated

Includes:Polypropylene screw closures

.

Specifications
Capacity (English)32 oz.
Closure Size53 mm
Closure TypeScrew
DescriptionFlask Erlenmeyer Screw Closure PMP 1000mL
No. per Case6
No. per Pack2
AutoclavableAutoclavable
Bottom ShapePlain
Capacity (Metric)1000 mL
DisposableReusable
GraduatedYes
MaterialPMP
Mouth StyleNarrow
SterilityNon-sterile
Unit SizePack of 2