Glutaric dialdehyde, 50 wt.% solution in water
Glutaric dialdehyde, 50 wt.% solution in water
Glutaric dialdehyde, 50 wt.% solution in water
Thermo Scientific Chemicals

Glutaric dialdehyde, 50 wt.% solution in water

CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
檢視全部
Glass bottle
產品號碼 410960025
價格 (TWD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS111-30-8
IUPAC Namepentanedial
Molecular FormulaC5H8O2
InChI KeySXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILESO=CCCCC=O
檢視更多
規格Specification Sheet規格表
Appearance (Form)Clear solution
Strength50 to 51 %
pH3.2 to 4.2
Methanol=<0.5 % (GC)
Color scale=<50 APHA
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only