Glutaric dialdehyde, 8% solution in water
Glutaric dialdehyde, 8% solution in water
Glutaric dialdehyde, 8% solution in water
Thermo Scientific Chemicals

Glutaric dialdehyde, 8% solution in water

CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
Have Questions?
Catalog NumberQuantity
410971000100 g
Catalog number 410971000
Price (USD)
153.65
Online exclusive
170.00
Save 16.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
153.65
Online exclusive
170.00
Save 16.35
Each
Chemical Identifiers
CAS111-30-8
IUPAC Namepentanedial
Molecular FormulaC5H8O2
InChI KeySXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILESO=CCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific gravity(20/20°C) 0.99 to 1.20
Appearance (Form)Clear solution
Titration with NaOH7.0 to 9.5 wt.%
Color scale=<50 APHA
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only