beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%
beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%
beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%
Thermo Scientific Chemicals

beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%

beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%, C3H7Na2O6P.5H2O, CAS Number-13408-09-8 | CAS: 13408-09-8 | C3H19Na2O11P | 308.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Glass bottle
Catalog number 410995000
Price (TWD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13408-09-8
IUPAC Name[(1,3-dihydroxypropan-2-yl)oxy]phosphonic acid pentahydrate disodium
Molecular FormulaC3H19Na2O11P
InChI KeyVWVLGHQKEJESPW-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Isomer=<2 % alpha-Isomer
Appearance (Form)Crystalline powder
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only