Glyoxylic acid, 50% in water
Glyoxylic acid, 50% in water
Glyoxylic acid, 50% in water
Glyoxylic acid, 50% in water
Thermo Scientific Chemicals

Glyoxylic acid, 50% in water

CAS: 298-12-4 | C2H2O3 | 74.035 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 411065000
Price (USD)
76.50
Each
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
76.50
Each
Add to cart
Chemical Identifiers
CAS298-12-4
IUPAC Name2-oxoacetic acid
Molecular FormulaC2H2O3
InChI KeyHHLFWLYXYJOTON-UHFFFAOYSA-N
SMILESOC(=O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Refractive index1.4130 to 1.4200 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Titration with NaOH>=50 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only