Guanosine, 99%
Guanosine, 99%
Guanosine, 99%
Thermo Scientific Chemicals

Guanosine, 99%

CAS: 118-00-3 | C10H13N5O5 | 283.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 411130250
Price (EUR)
62,65
Online exclusive
69,30
Save 6,65
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
62,65
Online exclusive
69,30
Save 6,65
Each
Chemical Identifiers
CAS118-00-3
IUPAC Name2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular FormulaC10H13N5O5
InChI KeyNYHBQMYGNKIUIF-UUOKFMHZSA-N
SMILESNC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=98.5 to 101.5 % (On dry substance)
Appearance (Form)Powder
Appearance (Color)White to light yellow
Infrared spectrumConforms
Loss on drying=<5 % (1 g, 105°C, 3 h)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only