Guanylurea sulfate, 97%
Guanylurea sulfate, 97%
Guanylurea sulfate, 97%
Thermo Scientific Chemicals

Guanylurea sulfate, 97%

CAS: 591-01-5 | C4H12N8O2·H2SO4 | 302.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 411141000
Price (USD)
376.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
376.00
Each
Chemical Identifiers
CAS591-01-5
IUPAC Namebis((diaminomethylidene)urea); sulfuric acid
Molecular FormulaC4H14N8O6S
InChI KeyIATXFPUBPMZBPH-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.NC(N)=NC(N)=O.NC(N)=NC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration with NaOH96.0 to 104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only