Heparin sodium
Heparin sodium
Heparin sodium
Thermo Scientific Chemicals

Heparin sodium

Heparin Sodium Salt, CAS # 9041-08-1, is derived from porcine intestinal mucosa and shows blood anti-coagulant activity. | CAS: 9041-08-1
数量:
1 g
5 g
パッケージ:
Glass Bottle
製品番号(カタログ番号) 411210010
価格(JPY)
-
見積もりを依頼する
数量:
1 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9041-08-1
MDL NumberMFCD00081689
Appearance (Color)White to almost white
Infrared spectrumConforms
Loss on drying=<8.0 % (1 g, 60°C, 3 h) (P2O5)
Protein=<0.5 % (dried substance)
UV(40mg in 10ml water) Nucleotidic impurities
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Heparin sodium salt displays anti-coagulant activity. It can also efficiently inhibit mesenchymal cell proliferation and promotes the proliferation of primary epithelial cells
  • This compound can bind to antithrombin III and inhibit coagulation proteases

Applications

  • In in vitro studies, it is used as the main component of serum-free media to prevent cell aggregation
  • Heparin sodium salt can also induce the growth of human embryonic stem cells in a defined serum-free medium, acting as a cofactor for a basic fibroblast growth factor (FGF-2)
  • It is also applied in laboratory studies related to stem cell differentiation and growth
  • In in vitro studies this compound exhibits cytotoxic action inducing cancer cell death by interfering with transcription factor activity
RUO – Research Use Only