Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile
Thermo Scientific™

Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile

Green features
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing, and storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeClosure Type
4116-0500500 mL45-430 mmVented
4113-0125125 mL38-430 mmSolid, non-vented
4113-0250250 mL38-430 mmSolid, non-vented
4113-0500500 mL45-430 mmSolid, non-vented
4113-10001000 mL45-430 mmSolid, non-vented
4113-20002000 mL45-430 mmSolid, non-vented
4113-28002800 mL70 mmSolid, non-vented
4116-0125125 mL38-430 mmVented
4116-0250250 mL38-430 mmVented
4116-10001000 mL45-430 mmVented
4116-20002000 mL45-430 mmVented
4116-28002800 mL70 mmVented
Catalog number 4116-0500
Price (CLP)
160.681
Case of 12
Capacity (Metric):
500 mL
Closure Size:
45-430 mm
Closure Type:
Vented
Price (CLP)
160.681
Case of 12

Save preparation time and avoid contamination risk by using Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing, and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.

Features

  • Sterile and single use to reduce contamination risk
  • BPA-free
  • Collapses when autoclaved to reduce bio hazardous waste
  • Graduated for quick volume assessment

Warranty: 90 days

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (English)16 oz.
Closure Size45-430 mm
Closure TypeVented
Green FeaturesACT Ecolabel (Silver)
No. per Case12
No. per Pack1
ShapeErlenmeyer
Surface CoatingNone
AutoclavableNot Autoclavable
Bottom ShapeBaffled
Capacity (Metric)500 mL
DisposableSingle-use
GraduatedYes
MaterialPETG
Mouth StyleNarrow
SterilitySterile
Unit SizeCase of 12