1-Hexanesulfonic acid, sodium salt hydrate, 98%, HPLC grade
1-Hexanesulfonic acid, sodium salt hydrate, 98%, HPLC grade
1-Hexanesulfonic acid, sodium salt hydrate, 98%, HPLC grade
Thermo Scientific Chemicals

1-Hexanesulfonic acid, sodium salt hydrate, 98%, HPLC grade

CAS: 2832-45-3 | C6H13NaO3S | 188.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 411601000
Price (USD)
999.65
Special offer
Online exclusive
Ends: 28-Dec-2026
1,176.00
Save 176.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
999.65
Special offer
Online exclusive
Ends: 28-Dec-2026
1,176.00
Save 176.35
Each
Chemical Identifiers
CAS2832-45-3
IUPAC Namesodium hexane-1-sulfonate
Molecular FormulaC6H13NaO3S
InChI KeyQWSZRRAAFHGKCH-UHFFFAOYSA-M
SMILES[Na+].CCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
UV(0.25 M in water) 1 cm cell vs HPLC-grade water
UVat 240 nm A: =<0.05
Appearance (Color)White
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only