Histamine, 97%
Histamine, 97%
Histamine, 97%
Thermo Scientific Chemicals

Histamine, 97%

CAS: 51-45-6 | C5H9N3 | 111.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 411710010
Price (EUR)
70,65
Online exclusive
78,30
Save 7,65 (10%)
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
70,65
Online exclusive
78,30
Save 7,65 (10%)
Each
Chemical Identifiers
CAS51-45-6
IUPAC Name2-(1H-imidazol-5-yl)ethan-1-amine
Molecular FormulaC5H9N3
InChI KeyNTYJJOPFIAHURM-UHFFFAOYSA-N
SMILESNCCC1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder and/or chunks
Infrared spectrumConforms
Appearance (Color)White to light yellow
Melting point81°C to 85°C
Titration with HClO4>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only