4-Hydrazinobenzenesulfonic acid, hemihydrate, 98%
4-Hydrazinobenzenesulfonic acid, hemihydrate, 98%
4-Hydrazinobenzenesulfonic acid, hemihydrate, 98%
Thermo Scientific Chemicals

4-Hydrazinobenzenesulfonic acid, hemihydrate, 98%

CAS: 854689-07-9 | C6H8N2O31/2H2O | 197.21 g/mol
数量:
50 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 411780500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS854689-07-9
IUPAC Name4-hydrazinylbenzene-1-sulfonic acid hydrate
Molecular FormulaC6H10N2O4S
InChI KeyOSOMMFVBTYFMBI-UHFFFAOYSA-N
SMILESO.NNC1=CC=C(C=C1)S(O)(=O)=O
さらに表示
Appearance (Color)Light yellow to light orange
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Loss on drying4 % to 5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only