Hydrogen peroxide, ACS reagent, 30 wt% solution in water, nonstabilized, Thermo Scientific Chemicals
Hydrogen peroxide, ACS reagent, 30 wt% solution in water, nonstabilized, Thermo Scientific Chemicals
Hydrogen peroxide, ACS reagent, 30 wt% solution in water, nonstabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrogen peroxide, ACS reagent, 30 wt% solution in water, nonstabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
411881000100gPlastic bottle
4118800252.5kgPlastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 411881000
Price (EUR)
24,65
Online exclusive
27,50
Save 2,85 (10%)
Each
Quantity:
100g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
24,65
Online exclusive
27,50
Save 2,85 (10%)
Each
Chemical Identifiers
CAS7722-84-1
IUPAC Nameperoxol
Molecular FormulaH2O2
InChI KeyMHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILESOO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear solution
Color scale=<10 APHA
Titration with KMnO429.0 to 32.0 %
Heavy metals (as Pb)=<1 ppm
Chloride (Cl)=<2 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only