4-Hydroxyphthalic acid, 98%
4-Hydroxyphthalic acid, 98%
4-Hydroxyphthalic acid, 98%
Thermo Scientific Chemicals

4-Hydroxyphthalic acid, 98%

CAS: 610-35-5 | C8H6O5 | 182.13 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
產品號碼 412130050
價格 (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS610-35-5
IUPAC Name4-hydroxybenzene-1,2-dicarboxylic acid
Molecular FormulaC8H6O5
InChI KeyMWRVRCAFWBBXTL-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(O)C=C1C(O)=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to light brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only