4,5-Imidazoledicarboxylic acid, 99%, Thermo Scientific Chemicals
4,5-Imidazoledicarboxylic acid, 99%, Thermo Scientific Chemicals
4,5-Imidazoledicarboxylic acid, 99%, Thermo Scientific Chemicals
Acros Organics

4,5-Imidazoledicarboxylic acid, 99%, Thermo Scientific Chemicals

CAS: 570-22-9 | Molecular Formula: C5H4N2O4 | Molecular Weight: 156.10 g/mol
Have Questions?
Catalog NumberQuantity
412250250
also known as 41225
25 g
Catalog number 412250250
also known as 41225
HSN 29332990
Price (INR)
25,594.00
Each
Quantity:
25 g
Request bulk or custom format
Price (INR)
25,594.00
Each
Chemical Identifiers
CAS570-22-9
IUPAC Name1H-imidazole-4,5-dicarboxylic acid
Molecular FormulaC5H4N2O4
InChI KeyZEVWQFWTGHFIDH-UHFFFAOYSA-N
SMILESOC(=O)C1=C(N=CN1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to yellow-brown
Appearance (Form)Crystalline powder
Titration excess NaOH>=98.5 %
Infrared spectrumConforms

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only