1-Iodooctane, 98%, stabilized
1-Iodooctane, 98%, stabilized
1-Iodooctane, 98%, stabilized
Thermo Scientific Chemicals

1-Iodooctane, 98%, stabilized

CAS: 629-27-6 | C8H17I | 240.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 412461000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS629-27-6
IUPAC Name1-iodooctane
Molecular FormulaC8H17I
InChI KeyUWLHSHAHTBJTBA-UHFFFAOYSA-N
SMILESCCCCCCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to amber
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4868 to 1.4888 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only