Isopropyl bromoacetate, 97%
Isopropyl bromoacetate, 97%
Isopropyl bromoacetate, 97%
Thermo Scientific Chemicals

Isopropyl bromoacetate, 97%

CAS: 29921-57-1 | C5H9BrO2 | 181.03 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
Packaging:
檢視全部
Glass bottle
產品號碼 412810250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS29921-57-1
IUPAC Namepropan-2-yl 2-bromoacetate
Molecular FormulaC5H9BrO2
InChI KeyJCWLEWKPXYZHGQ-UHFFFAOYSA-N
SMILESCC(C)OC(=O)CBr
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4440 to 1.4460 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only