Isopropyl myristate, 96%
Isopropyl myristate, 96%
Isopropyl myristate, 96%
Thermo Scientific Chemicals

Isopropyl myristate, 96%

CAS: 110-27-0 | C17H34O2 | 270.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Catalog number 412851000
Price (USD)
37.90
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
37.90
Each
Chemical Identifiers
CAS110-27-0
IUPAC Namepropan-2-yl tetradecanoate
Molecular FormulaC17H34O2
InChI KeyAXISYYRBXTVTFY-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCC(=O)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<30 APHA
Water=<0.1 %
Refractive index1.4340 to 1.4360 (20°C, 589 nm)
Iodine value=<1 g I2/100 g
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only