Thermo Scientific Chemicals

DL-Lactic acid, 85+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 412965000
500 g, EcoSafPak™ Glass Bottle, Each
Thermo Scientific Chemicals

DL-Lactic acid, 85+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 412965000
500 g, EcoSafPak™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 412965000
Price (USD)
Price: 71.60
Online price: 60.65
Your price:
Quantity
-

Chemical Identifiers

CAS
50-21-5
IUPAC Name
2-hydroxypropanoic acid
Molecular Formula
C3H6O3
InChI Key
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
SMILES
CC(O)C(O)=O
Heavy metals (as Pb)
=<5 ppm
Chloride (Cl)
=<10 ppm
Residue after ignition
=<0.02 %
Appearance (Color)
Clear colorless
Titration excess NaOH
85.0 to 90.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text