Capit-All Flex  Cap Driver Cassettes
Thermo Scientific™
Capit-All Flex Cap Driver Cassettes
The Thermo Scientific Capit-All Flex Cap Driver Cassettes are compatible with both Matrix and Nunc tubes in 96 or 48 formats, and Azenta FluidX tubes in 96 format. It is recommended to be replaced after 10,000 decapping cycles.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTubos de almacenamiento compatibles
4130-FDX-EAzenta 96 Format External Thread
4130-FDX-IAzenta 96 Format Internal Thread
4130-MAT-NUNMatrix and Nunc tubes in 96 or 48 formats
Número de catálogo 4130-FDX-E
Precio (CLP)
-
Tubos de almacenamiento compatibles:
Azenta 96 Format External Thread
The Thermo Scientific Capit-All Flex Cap Driver Cassettes are compatible with both Matrix and Nunc tubes in 96 or 48 formats, and Azenta FluidX tubes in 96 format. It is recommended to be replaced after 10,000 decapping cycles.
  • Cap driver cassette compatible with Thermo Scientific Capit-All Flex Automated Decapper (P/N 4120-FLEX)
  • Tube compatibility: Matrix and Nunc cryotubes in 96 and 48 formats; Azenta FluidX tubes with internal threads in 96 format
  • Replace after 10,000 cycles
Especificaciones
Tubos de almacenamiento compatiblesAzenta 96 Format External Thread
Formato de destapado96-Channel
TipoDecapping Head
Unit SizeEach