Thermo Scientific™

Forma™ Series 3 Water Jacketed CO2 Incubator, 184L

Catalog number: 4130TS
Have Questions?
Thermo Scientific™

Forma™ Series 3 Water Jacketed CO2 Incubator, 184L

Catalog number: 4130TS
Have Questions?
The Thermo Scientific™ Forma Series 3 Water Jacketed CO2 Incubator includes reliable water jacket technology with effortless Thermofisher Scientific™ iCAN™ touchscreen. Dependable temperature stability and HEPA clean room air purity are paired with the simplest way to control and monitor your incubator, ensuring protection for your important cell cultures, a combination that’s truly hard to beat.
 
Catalog Number
4130TS
Unit Size
Each
Electrical Requirements
115 V, 50/60 Hz
CO2 Sensor Technology
TC Sensor
Chamber Material
Polished Stainless Steel
Oxygen Control
1 to 20%
Price (USD)
Full specifications
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle Tri-gas 184L incubator, 1-20%
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyTC Sensor
LightingLED
MaterialStainless Steel
O2 Concentration Range1 to 20%
Oxygen Control1 to 20%
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements115 V, 50/60 Hz
Voltage115 V
Unit SizeEach
Catalog Number
Unit Size
Each
Electrical Requirements
115 V, 50/60 Hz
CO2 Sensor Technology
IR Sensor
Chamber Material
Polished Stainless Steel
Oxygen Control
None
Price (USD)
Full specifications
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle 184L incubator
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyIR Sensor
LightingLED
MaterialStainless Steel
Oxygen ControlNone
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements115 V, 50/60 Hz
Voltage115 V
Unit SizeEach
Catalog Number
4121TS
also known as 4121
Unit Size
Each
Electrical Requirements
230 V, 50 Hz
CO2 Sensor Technology
IR Sensor
Chamber Material
Polished Stainless Steel
Oxygen Control
None
Price (USD)
Full specifications
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle 184L incubator
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyIR Sensor
LightingLED
MaterialStainless Steel
Oxygen ControlNone
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements230 V, 50 Hz
Voltage230 V
Unit SizeEach
Catalog Number
4140TS
also known as 4140
Unit Size
Each
Electrical Requirements
115 V, 50/60 Hz
CO2 Sensor Technology
IR Sensor
Chamber Material
Polished Stainless Steel
Oxygen Control
1 to 20%
Price (USD)
Full specifications
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle Tri-gas 184L incubator, 1-20%
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyIR Sensor
LightingLED
MaterialStainless Steel
O2 Concentration Range1 to 20%
Oxygen Control1 to 20%
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements115 V, 50/60 Hz
Voltage115 V
Unit SizeEach
Catalog Number
Unit Size
Each
Electrical Requirements
230 V, 50 Hz
CO2 Sensor Technology
IR Sensor
Chamber Material
Polished Stainless Steel
Oxygen Control
1 to 20%
Price (USD)
Full specifications
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle Tri-gas 184L incubator, 1-20%
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyIR Sensor
LightingLED
MaterialStainless Steel
O2 Concentration Range1 to 20%
Oxygen Control1 to 20%
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements230 V, 50 Hz
Voltage230 V
Unit SizeEach
Catalog Number
Unit Size
Each
Electrical Requirements
115 V, 50/60 Hz
CO2 Sensor Technology
TC Sensor
Chamber Material
Polished Stainless Steel
Oxygen Control
None
Price (USD)
Full specifications
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle 184L incubator
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyTC Sensor
LightingLED
MaterialStainless Steel
Oxygen ControlNone
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements115 V, 50/60 Hz
Voltage115 V
Unit SizeEach
Catalog Number
4111TS
also known as 4111
Unit Size
Each
Electrical Requirements
230 V, 50/60 Hz
CO2 Sensor Technology
TC Sensor
Chamber Material
Polished Stainless Steel
Oxygen Control
None
Price (USD)
Full specifications
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle 184L incubator
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyTC Sensor
LightingLED
MaterialStainless Steel
Oxygen ControlNone
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Unit SizeEach
Catalog Number
Unit Size
Each
Electrical Requirements
230 V, 50/60 Hz
CO2 Sensor Technology
TC Sensor
Chamber Material
Polished Stainless Steel
Oxygen Control
1 to 20%
Price (USD)
Full specifications
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle Tri-gas 184L incubator, 1-20%
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyTC Sensor
LightingLED
MaterialStainless Steel
Oxygen Control1 to 20%
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Unit SizeEach
Showing 8 of 8
Catalog NumberSpecificationsUnit SizeElectrical RequirementsCO2 Sensor TechnologyChamber MaterialOxygen ControlPrice (USD)
4130TSFull specifications
Each115 V, 50/60 HzTC SensorPolished Stainless Steel1 to 20%Request A Quote
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle Tri-gas 184L incubator, 1-20%
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyTC Sensor
LightingLED
MaterialStainless Steel
O2 Concentration Range1 to 20%
Oxygen Control1 to 20%
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements115 V, 50/60 Hz
Voltage115 V
Unit SizeEach
4120Full specifications
Each115 V, 50/60 HzIR SensorPolished Stainless SteelNoneRequest A Quote
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle 184L incubator
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyIR Sensor
LightingLED
MaterialStainless Steel
Oxygen ControlNone
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements115 V, 50/60 Hz
Voltage115 V
Unit SizeEach
4121TS
also known as 4121
Full specifications
Each230 V, 50 HzIR SensorPolished Stainless SteelNoneRequest A Quote
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle 184L incubator
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyIR Sensor
LightingLED
MaterialStainless Steel
Oxygen ControlNone
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements230 V, 50 Hz
Voltage230 V
Unit SizeEach
4140TS
also known as 4140
Full specifications
Each115 V, 50/60 HzIR SensorPolished Stainless Steel1 to 20%Request A Quote
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle Tri-gas 184L incubator, 1-20%
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyIR Sensor
LightingLED
MaterialStainless Steel
O2 Concentration Range1 to 20%
Oxygen Control1 to 20%
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements115 V, 50/60 Hz
Voltage115 V
Unit SizeEach
4141Full specifications
Each230 V, 50 HzIR SensorPolished Stainless Steel1 to 20%Request A Quote
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle Tri-gas 184L incubator, 1-20%
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyIR Sensor
LightingLED
MaterialStainless Steel
O2 Concentration Range1 to 20%
Oxygen Control1 to 20%
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements230 V, 50 Hz
Voltage230 V
Unit SizeEach
4110Full specifications
Each115 V, 50/60 HzTC SensorPolished Stainless SteelNoneRequest A Quote
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle 184L incubator
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyTC Sensor
LightingLED
MaterialStainless Steel
Oxygen ControlNone
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements115 V, 50/60 Hz
Voltage115 V
Unit SizeEach
4111TS
also known as 4111
Full specifications
Each230 V, 50/60 HzTC SensorPolished Stainless SteelNoneRequest A Quote
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle 184L incubator
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyTC Sensor
LightingLED
MaterialStainless Steel
Oxygen ControlNone
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Unit SizeEach
4131Full specifications
Each230 V, 50/60 HzTC SensorPolished Stainless Steel1 to 20%Request A Quote
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Data OutputsUSB, and optional 4-20mA
DescriptionSingle Tri-gas 184L incubator, 1-20%
Capacity (Metric)184 L
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyTC Sensor
LightingLED
MaterialStainless Steel
Oxygen Control1 to 20%
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Dimensions (D x W x H) Interior20.0 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Dimensions (L x W x H)25 x 26 x 39.5 in. (63.5 x 66 x 100.3 cm)
Weight (English)365 lb.
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Unit SizeEach
Showing 8 of 8
iCAN touchscreen interface
  • Available in various languages including Japanese and Mandarin
  • Easy-to-use on-screen menu prompts
  • On-board error and data logging for monitoring performance, interactions and alarm conditions
  • Bright, door-mounted touchscreen interface provides complete data visibility to monitor all incubator interaction
Temperature stability with Water Jacket design
Unique triple-wall construction delivers optimal temperature uniformity. Provides ultimate thermal stability for protection for your valuable cell cultures against ambient temperature swings and unexpected power outages.
 
Protection with HEPA air filtration
Minimize the risk of airborne contaminants entering the incubator from multiple door openings. In-chamber HEPA filtration reaches ISO 5 clean room air quality in 5 minutes by filtering the chamber air every 60 seconds.
 
Enhanced flexibility with complete O2 control
Many cell cultures thrive best in CO2 incubators with controlled levels of oxygen. Select an O2 option to simulate physiological or hypoxic environments in a range of 1 to 20%. A dedicated O2 display set point and control allow for accurate monitoring.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text