Lithium hydroxide monohydrate, 98+%, ACS reagent
Lithium hydroxide monohydrate, 98+%, ACS reagent
Lithium hydroxide monohydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Lithium hydroxide monohydrate, 98+%, ACS reagent

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 413321000
Price (EUR)
52,90
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
52,90
Each
Chemical Identifiers
CAS1310-66-3
IUPAC Namelithium(1+) hydrate hydroxide
Molecular FormulaH3LiO2
InChI KeyGLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES[Li+].O.[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HCl>=98.0 %
Heavy metals (ICP-OES)=<0.002 %
Sulfate (SO4)=<0.05 %
Lithium carbonate=<2.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only