DL-Lysine monohydrochloride, 99%
DL-Lysine monohydrochloride, 99%
DL-Lysine monohydrochloride, 99%
Thermo Scientific Chemicals

DL-Lysine monohydrochloride, 99%

CAS: 70-53-1 | C6H15ClN2O2 | 182.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Packaging:
Glass Bottle
Catalog number 413371000
Price (TWD)
-
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS70-53-1
IUPAC Name2,6-diaminohexanoic acid hydrochloride
Molecular FormulaC6H15ClN2O2
InChI KeyBVHLGVCQOALMSV-UHFFFAOYSA-N
SMILESCl.NCCCCC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific optical rotation-1° to +1° (20°C, 589 nm) (c=1, water)
Appearance (Form)Powder
Appearance (Color)White to off-white
Titration with HClO498.5 to 101.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only