Mercury(II) sulfate, ACS reagent
Mercury(II) sulfate, ACS reagent
Mercury(II) sulfate, ACS reagent
Thermo Scientific Chemicals

Mercury(II) sulfate, ACS reagent

CAS: 7783-35-9 | HgO4S | 296.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 413655000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7783-35-9
IUPAC Namemercury(2+) sulfate
Molecular FormulaHgO4S
InChI KeyDOBUSJIVSSJEDA-UHFFFAOYSA-L
SMILES[Hg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to off-white
Titration with NH4SCN>=98.0 %
Nitrate (NO3)Passes test (limit about 0.003%)
Chloride (Cl)=<30 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only