Metanil Yellow (Tech.), 85%
Metanil Yellow (Tech.), 85%
Metanil Yellow (Tech.), 85%
Thermo Scientific Chemicals

Metanil Yellow (Tech.), 85%

CAS: 587-98-4 | C18H14N3NaO3S | 375.378 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
Catalog number 413722500
Price (USD)
95.65
Online exclusive
106.00
Save 10.35
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
95.65
Online exclusive
106.00
Save 10.35
Each
Chemical Identifiers
CAS587-98-4
IUPAC Namesodium 3-{2-[4-(phenylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
Molecular FormulaC18H14N3NaO3S
InChI KeyNYGZLYXAPMMJTE-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)C1=CC=CC(=C1)N=NC1=CC=C(NC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
UV Lambda max413 to 419 nm (in methanol)
Appearance (Color)Golden to orange
Visual transition intervalfrom pH 1.2 (red) to 2.3 (yellow)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only