N-Methyldiphenylamine, 97%
N-Methyldiphenylamine, 97%
N-Methyldiphenylamine, 97%
Thermo Scientific Chemicals

N-Methyldiphenylamine, 97%

CAS: 552-82-9 | C13H13N | 183.25 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 414210050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS552-82-9
IUPAC NameN-methyl-N-phenylaniline
Molecular FormulaC13H13N
InChI KeyDYFFAVRFJWYYQO-UHFFFAOYSA-N
SMILESCN(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear yellow to orange-red
Refractive index1.6215 to 1.6235 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only